Alfa Aesar™ Tellurium powder, -18+60 mesh, Puratronic™, 99.999% (metals basis)

Manufacturer: Alfa Aesar™ 4396018

Quantity 50g

 View more versions of this product

Catalog No. AA4396018

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13494-80-9
    MDL Number MFCD00134062
    UN Number UN3288
    Chemical Name or Material Tellurium powder
    Grade Puratronic™

    Specifications

    Quantity 50g
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 13494-80-9
    TSCA TSCA
    MDL Number MFCD00134062
    Odor Odorless
    Physical Form Powder
    UN Number UN3288
    Health Hazard 2 GHS H Statement
    H301-H332-H315-H319-H335
    Toxic if swallowed.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
    Chemical Name or Material Tellurium powder
    Assay 99.999%, (metals basis)
    Grade Puratronic™

    GHS H Statement
    H301-H332-H315-H319-H335
    Toxic if swallowed.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit