Tellurium Powder, -60 mesh, 99.999% (Metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 1107509

Quantity 10g

 View more versions of this product

Catalog No. AA1107509

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13494-80-9
    Physical Form Powder
    Assay Percent Range 99.999% (Metals basis)
    MDL Number MFCD00134062
    Chemical Name or Material Tellurium

    Specifications

    Quantity 10g
    Color Gray
    DOT Information DOT Class: 6.1, Packaging Group: III
    EINECS Number 236-813-4
    Physical Form Powder
    Odor Odorless
    Assay Percent Range 99.999% (Metals basis)
    Appearance Gray powder
    MDL Number MFCD00134062
    CAS 13494-80-9
    Health Hazard 1 H301-H332-H315-H319-H335; Toxic if swallowed. Harmful if inhaled. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
    Chemical Name or Material Tellurium

    H301-H332-H315-H319-H335; Toxic if swallowed. Harmful if inhaled. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit