Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tellurium shot, 99.9999% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 4221322

Quantity 100g

 View more versions of this product

Catalog No. AA4221322


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

13494-80-9
99.9999% (Metals basis)
Tellurium
Shot, 2-5mm (0.08-0.20 in.) dia.
MFCD00134062

Specifications

13494-80-9
99.9999% (Metals basis)
MFCD00134062
Odorless
100g
Shot, 2-5mm (0.08-0.20 in.) dia.
Silvery-white shot
6.24g/cm3
Silver-white
Tellurium

Safety and Handling

SDS & Safety Documents

H300-H332: Fatal if swallowed. Harmful if inhaled.

EINECSNumber : 236-813-4

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit