Telmisartan, Tocris™

Manufacturer: R&D Systems 5139/50

Quantity 50mg

 View more versions of this product

Catalog No. 51-395-0R

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 144701-48-4
    Physical Form Solid
    Assay Percent Range >99%
    Molecular Formula C33H30N4O2
    Formula Weight 514.62
    Chemical Name or Material Telmisartan

    Specifications

    Physical Form Solid
    Formula Weight 514.62
    Color White
    Quantity 50mg
    Assay Percent Range >99%
    Molecular Formula C33H30N4O2
    CAS 144701-48-4
    Recommended Storage Store at RT
    Chemical Name or Material Telmisartan

    Store at RT

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit