Learn More
TEM1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 181601AP
Catalog No. 50-173-2023
Add to cart
Description
Description
CD248 (CD248 Molecule) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene include calcium ion binding and extracellular matrix binding. An important paralog of this gene is CD93. CD248 may play a role in tumor angiogenesis.
Specifications
Specifications
TEM1
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q91V98, Q9HCU0
CD164L1, CD248, CD248 molecule, endosialin, Endosialin, TEM1, Tumor endothelial marker 1
Rabbit
IgG
150 μL
-20°C
Primary
293669, 57124, 70445
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
CD248
Liquid
Cd248
TEM1 Fusion Protein Ag12990
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat