Learn More
TEM5/GPR124 Mouse anti-Human, Alexa Fluor 488, Clone: 499716, R&D Systems™
Mouse MonoclonalAntibody
Manufacturer:  R&D Systems FAB8896G100UG
Catalog No. FAB8896G100
Add to cart
This item is not returnable. View return policy
Description
Description
TEM5/GPR124 Monoclonal antibody specifically detects TEM5/GPR124 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
TEM5/GPR124
Monoclonal
Alexa Fluor 488
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
DKFZp434C211, DKFZp434J0911, G protein-coupled receptor 124, GPR124, G-protein coupled receptor 124, KIAA1531, KIAA1531FLJ14390, TEM5Tumor endothelial marker 5
Chinese hamster ovary cell line CHO-derived recombinant human TEM5/GPR124 Met1-Leu699 Accession # Q96PE1
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Detects human TEM5/GPR124 in direct ELISAs.
Flow Cytometry
499716
Flow Cytometry 0.25-1 ug/10^6 cells
Q96PE1
Mouse
IgG2b
RUO
Primary
25960
Human
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)