Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TEM5/GPR124 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB88961SP
This item is not returnable.
View return policy
Description
TEM5/GPR124 Monoclonal antibody specifically detects TEM5/GPR124 in Human samples. It is validated for nanSpecifications
TEM5/GPR124 | |
499708 | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
25960 | |
Human | |
Purified |
Monoclonal | |
Unconjugated | |
DKFZp434C211, DKFZp434J0911, G protein-coupled receptor 124, G-protein coupled receptor 124, KIAA1531FLJ14390, TEM5Tumor endothelial marker 5 | |
Chinese hamster ovary cell line CHO-derived recombinant human TEM5/GPR124, Met1-Leu699, Accession # Q96PE1 | |
25 μg | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction