Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ TEM7/PLXDC1 Antibody Pair [HRP]

Mouse Monoclonal Antibody Pair

Manufacturer:  Novus Biologicals™ NBP2793635PLATES

 View more versions of this product

Catalog No. NB009766


Add to cart

Description

Description

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human TEM7/PLXDC1.
Specifications

Specifications

156.25 pg/mL
Sandwich ELISA
  • Detection Antibody: 0.5 mg/mL mouse anti-TEM7/PLXDC1 monoclonal antibody conjugated to horseradish-peroxidase (HRP).
  • Capture Antibody: 0.3 mg/mL of mouse anti-TEM7/PLXDC1 monoclonal antibody.
  • Standard: Each vial contains 270 ng of recombinant TEM7/PLXDC1.
  • RUO
    This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human TEM7/PLXDC1.
    TEM7/PLXDC1
    Angiogenesis
    Human
    PLXDC1
    1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 10 ng/mL is recommended.
    HRP
    57125
    Antibody Pairs
    156.25-10000 pg/mL
    Storage is content dependent.
    Mouse
    5 Plates
    Sandwich ELISA
    FLJ45632, plexin domain containing 1,2410003I07Rik, plexin domain-containing protein 1, TEM3FLJ36270, TEM7DKFZp686F0937, Tumor endothelial marker 3, Tumor endothelial marker 7
    SDS
    Documents
    Provide Content Correction

    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit