TEMED, OmniPur™, Calbiochem™, EMD Millipore

Free radical initiator and stabilizer used in acrylamide polymerization for PAGE gels

Manufacturer: EMD Millipore 892025ML

Quantity 25mL

 View more versions of this product

Catalog No. 89-202-5ML

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 110-18-9
    Physical Form Liquid
    Molecular Formula C6H16N2
    Formula Weight 116.2
    Color Clear
    Protease Not detected
    RNase Not detected
    DNase Not detected
    Chemical Name or Material TEMED
    Grade Molecular biology grade

    Specifications

    Formula Weight 116.2
    Boiling Point 121°C
    Melting Point −55°C
    Quantity 25mL
    Synonym N,N,N',N'-Tetramethylethylenediamine
    DNase Not detected
    Molecular Formula C6H16N2
    UN Number UN 2372
    Protease Not detected
    RNase Not detected
    Color Clear
    Flash Point 20.85°C (69.5°F)
    Recommended Storage Store in accordance with local regulations. Store in a segregated and approved area. Store in original container, protected from direct sunlight. Eliminate all ignition sources. Separate from oxidizing materials. Keep container tightly closed and sealed until ready for use. Containers that have been opened must be carefully resealed and kept upright to prevent leakage.
    DOT Information DOT Class: 3, Packaging Group: II
    CAS 110-18-9
    Odor Amine-like
    Physical Form Liquid
    Solubility Information Water: Soluble
    Chemical Name or Material TEMED
    Grade Molecular biology grade

    Store in accordance with local regulations. Store in a segregated and approved area. Store in original container, protected from direct sunlight. Eliminate all ignition sources. Separate from oxidizing materials. Keep container tightly closed and sealed until ready for use. Containers that have been opened must be carefully resealed and kept upright to prevent leakage.

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit