Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ TEMED, OmniPur™, Calbiochem™,

Catalog Number 8930100ML
Click to view available options
Quantity:
25 mL
100 mL
110-18-9
C6H16N2
116.208
KWYHDKDOAIKMQN-UHFFFAOYSA-N
temed, n,n,n',n'-tetramethylethylenediamine, tmeda, 1,2-bis dimethylamino ethane, tetramethylethylenediamine, tetramethyldiaminoethane, tetrameen, propamine d, n1,n1,n2,n2-tetramethylethane-1,2-diamine, 1,2-ethanediamine, n,n,n',n'-tetramethyl
8037
CHEBI:32850
N,N,N',N'-tetramethylethane-1,2-diamine
CN(C)CCN(C)C
110-18-9
C6H16N2
116.208
KWYHDKDOAIKMQN-UHFFFAOYSA-N
temed, n,n,n',n'-tetramethylethylenediamine, tmeda, 1,2-bis dimethylamino ethane, tetramethylethylenediamine, tetramethyldiaminoethane, tetrameen, propamine d, n1,n1,n2,n2-tetramethylethane-1,2-diamine, 1,2-ethanediamine, n,n,n',n'-tetramethyl
8037
CHEBI:32850
N,N,N',N'-tetramethylethane-1,2-diamine
CN(C)CCN(C)C

Free radical initiator and stabilizer used in acrylamide polymerization for PAGE gels

Melting Point -55°C
Boiling Point 121°C
Flash Point 20.85°C (69.5°F)
Odor Amine-like
Quantity 100 mL
UN Number UN 2372
Solubility Information Water: Soluble
Formula Weight 116.2
Physical Form Liquid
Chemical Name or Material TEMED
Grade Molecular biology grade
Show More Show Less
DOTInformation DOT Class: 3, Packaging Group: II
Recommended Storage Store in accordance with local regulations. Store in a segregated and approved area. Store in original container, protected from direct sunlight. Eliminate all ignition sources. Separate from oxidizing materials. Keep container tightly closed and sealed until ready for use. Containers that have been opened must be carefully resealed and kept upright to prevent leakage.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit