TEMED, Ultrapure Bioreagent, J.T.Baker™

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

Manufacturer: J.T.Baker 409800

Packaging Narrow Mouth Amber Glass Bottle
Quantity 5mL

 View more versions of this product

Catalog No. 02-004-459

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 110-18-9
    Molecular Formula (CH3)2NCH2CH2N(CH3)2
    Formula Weight 116.21
    Chemical Name or Material TEMED, Catalyst for Polyacrylamide Gels
    Grade Ultrapure Bioreagent

    Specifications

    Formula Weight 116.21
    Molecular Formula (CH3)2NCH2CH2N(CH3)2
    Quantity 5mL
    CAS 110-18-9
    Specific Gravity 1L = 0.78kg
    Flash Point 10.00°C
    Packaging Narrow Mouth Amber Glass Bottle
    Chemical Name or Material TEMED, Catalyst for Polyacrylamide Gels
    Grade Ultrapure Bioreagent

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit