Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TEMED, Ultrapure Bioreagent, J.T.Baker™

Click to view available options
Quantity:
50mL
100mL
Packaging:
Amber Glass Bottle
Narrow Mouth Amber Glass Bottle
110-18-9
C6H16N2
116.208
KWYHDKDOAIKMQN-UHFFFAOYSA-N
temed,n,n,n',n'-tetramethylethylenediamine,tmeda,1,2-bis dimethylamino ethane,tetramethylethylenediamine,tetramethyldiaminoethane,tetrameen,propamine d,n1,n1,n2,n2-tetramethylethane-1,2-diamine,1,2-ethanediamine, n,n,n',n'-tetramethyl
8037
CHEBI:32850
N,N,N',N'-tetramethylethane-1,2-diamine
CN(C)CCN(C)C
110-18-9
C6H16N2
116.208
KWYHDKDOAIKMQN-UHFFFAOYSA-N
temed,n,n,n',n'-tetramethylethylenediamine,tmeda,1,2-bis dimethylamino ethane,tetramethylethylenediamine,tetramethyldiaminoethane,tetrameen,propamine d,n1,n1,n2,n2-tetramethylethane-1,2-diamine,1,2-ethanediamine, n,n,n',n'-tetramethyl
8037
CHEBI:32850
N,N,N',N'-tetramethylethane-1,2-diamine
CN(C)CCN(C)C

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

Flash Point 10.00°C
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 0.78kg
Quantity 100mL
Formula Weight 116.21
Grade Ultrapure Bioreagent
Chemical Name or Material TEMED, Catalyst for Polyacrylamide Gels