Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TEMED, Ultrapure Bioreagent, J.T.Baker™

missing translation for 'orderingAttributeHoverText'
Quantity:
100mL
50mL
Packaging:
Amber Glass Bottle
Narrow Mouth Amber Glass Bottle

Chemical Identifiers

110-18-9
C6H16N2
116.208
KWYHDKDOAIKMQN-UHFFFAOYSA-N
8037
CHEBI:32850

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

CAS 110-18-9
Molecular Formula C6H16N2
InChI Key KWYHDKDOAIKMQN-UHFFFAOYSA-N
SMILES CN(C)CCN(C)C
Packaging Amber Glass Bottle
IUPAC Name N,N,N',N'-tetramethylethane-1,2-diamine
Molecular Weight (g/mol) 116.208
PubChem CID 8037
ChEBI CHEBI:32850
Formula Weight 116.21
Specific Gravity 1L = 0.78kg
Flash Point 10.00°C
Quantity 50mL
Chemical Name or Material TEMED, Catalyst for Polyacrylamide Gels
Grade Ultrapure Bioreagent
Show More Show Less