Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Temozolomide, Tocris Bioscience™

Catalog Number p-4784673
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
85622-93-1
C6H6N6O2
194.154
BPEGJWRSRHCHSN-UHFFFAOYSA-N
temozolomide, methazolastone, temodal, temodar, temozolamide, temozolomidum, 3-methyl-4-oxo-3,4-dihydroimidazo 5,1-d 1,2,3,5 tetrazine-8-carboxamide, temozolomidum latin, temozolodida spanish, temozolodida
5394
CHEBI:72564
3-methyl-4-oxoimidazo[5,1-d][1,2,3,5]tetrazine-8-carboxamide
CN1C(=O)N2C=NC(=C2N=N1)C(=O)N
This item is not returnable. View return policy
85622-93-1
C6H6N6O2
194.154
BPEGJWRSRHCHSN-UHFFFAOYSA-N
temozolomide, methazolastone, temodal, temodar, temozolamide, temozolomidum, 3-methyl-4-oxo-3,4-dihydroimidazo 5,1-d 1,2,3,5 tetrazine-8-carboxamide, temozolomidum latin, temozolodida spanish, temozolodida
5394
CHEBI:72564
3-methyl-4-oxoimidazo[5,1-d][1,2,3,5]tetrazine-8-carboxamide
CN1C(=O)N2C=NC(=C2N=N1)C(=O)N

DNA-methylating antitumor agent

DNA methylating, chemotherapeutic agent. Displays antitumor activity against a board spectrum of tumors, including leukemias, lymphomas and solid tumors (IC50 = 5.0 μM for cytotoxicity against mouse TLX5 lymphoma cells). Induces autophagy in malignant glioma cells. Exhibits enhanced activity when used in combination with CHC (Cat No. 5029).
Quantity 10 mg
Formula Weight 194.15
Percent Purity >99%
Chemical Name or Material Temozolomide
Recommended Storage Store at Room Temperature
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit