Learn More
Cole-Parmer MadgeTech ULTRASHOCK-50EB Temp/RH/Pressure/Shock Logger  DFS Item
Manufacturer:  Cole-Parmer 1800303
The Shock/Acceleration series are battery powered, stand alone three -axis shock recorders. Each data logger measures and records shock as the peak acceleration levels over the user defined interval. It is specifically designed for documenting dynamic environments, such as moving vehicles, trucks, containers, and ships. The device is also valuable in characterizing environments, such as production and assembly lines of delicate equipment, IC fabrication, communications and computer components. This is an all-in-one compact, portable, easy to use device that will measure and record thousands of measurements per channel. The storage medium is non-volatile, solid-state memory providing maximum data security even if the battery becomes discharged. The device can be started and stopped directly from your computer and its small size allows it to fit almost anywhere. Data retrieval is quick and easy. Simply put it into an empty COM port and our user-friendly software does the rest.
Catalog No. 50-631-561