Learn More
Thermo Scientific™ Temperature Chart Recorders
Record seven-day history on 6 in. charts using Thermo Scientific™ Temperature Chart Recorders, for use with most freezers and refrigerators.
Manufacturer:  Thermo Scientific™ 63837
 View more versions of this product
Catalog No. 13-990-220
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Part Not Found
Specifications
Description
Record seven-day history on 6 in. charts using Thermo Scientific™ Temperature Chart Recorders, for use with most freezers and refrigerators. These recorders are ideal when ultra-low temperature freezers need to meet validation and regulatory requirements.
Features:
  • Seven-day chart recorders record temperatures from -100° to 0°C (-40° to +38° on 6183-7)
  • 6 in. (15.2cm) chart
  • Use pressure-sensitive paper, require no inking
  • Freestanding model is customer installed and must be installed by a qualified professional
  • Panel-mounted model is factory installed; order when purchasing refrigerator or freezer
Ordering Information:
Available in range of voltages; specify voltage when ordering.
Includes:
Pack of 50 charts
Description & Specifications
Specifications
General-Purpose, Chromatography Refrigerators
6 in.
Free standing; Customer installed
Customer-Installed,Free-Standing,Freestanding, customer installed
Seven-day chart recorders
15.2cm
-100° to 0°C,-100° to 0°C
General-Purpose, Chromatography Refrigerators