Tempshield™ Cryo-Gloves™ Get IT Item

Breathable, stay-dry lining

Manufacturer: Tempshield™ MAM

 View more versions of this product

Catalog No. 11-394-305

  Check Availability
Add to Cart

Alert

Not for immersion in liquid nitrogen or for use near open flame or in hazardous areas where ignition could occur.


    Specifications

    For Use With (Application) superior thermal protection when working with ultra low temperature freezers, blast freezers, dry ice and in coldrooms, also for use with Liquefied Natural Gas, liquid helium and liquid oxygen in enclosed systems
    Length (English) 13.5 to 15.25 in.
    Length (Metric) 34.5 to 39cm
    Temperature (English) Max. -321°F
    Size Medium
    Style Cryo-Gloves with Mid-Arm Length
    Temperature (Metric) Max -196°C
    Color Blue

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit