Learn More
Tempshield™ Cryo-Gloves™ Blue Shoulder-Length Cryogenic Gloves
SDP
State-of-the art materials and multi-layered construction provide cryogenic protection for ultra-cold applications down to -196°C (-320°F).
Supplier:  Tempshield™ SHXL
 View more versions of this product
Catalog No. 19-159178
Add to Cart
Alert:
Not for immersion in liquid nitrogen, for use near open flame, or in hazardous areas where ignition could occur.
This item is not returnable. View return policy
Description
Description
State-of-the art materials and multi-layered construction provides cryogenic protection for ultra-cold applications down to -196°C (-320°F).
  • Cryogenic protection for ultra-cold applications down to -196°C (-320°F)
  • State-of-the art materials and multi-layered construction allow for a maximum level of thermal protection, flexibility, and dexterity
  • High performance, thermal lining wicks moisture away from hands to maintain comfort over extended periods of wear
  • Ideal for applications where function is important and safety is critical
  • Not intended for immersion in liquid nitrogen or other cryogenic liquids
Specifications
Specifications
EN 420, EN 511
Blue
Cryogenic Gloves
Yes
Cryo-Gloves™
-196°C
62 to 69.5 cm
Shoulder Length
Specific
X-Large
Dry Ice Handling, Low/Ultra-Low Temperature Freezers, and Closed Cryogenic Systems
-320°F
24.5 to 27.3 in.
Product Suggestions
Product Suggestions