Learn More
Tempshield™ Pink Cryo-Gloves™
Manufacturer:  Tempshield™ PSHS
 View more versions of this product
Catalog No. 19-230-273
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Fabrics, Multi-layered Insulated Construction
Fabric
8,Small
Pink
62.5 to 68.5cm
LNG (Liquefied Natural Gas), Liquid Helium, and Liquid Oxygen in enclosed systems, ultra-low temperature freezers, blast freezers, dry ice, and in coldrooms
Thermal
-259°F
24.75 to 27 in.