Tempshield™ Pink Cryo-Gloves™
Manufacturer: Tempshield™ PSHXL
 View more versions of this product
Catalog No. 19-230-276
  Check Availability
Add to Cart
    Specifications
    For Use With (Application) LNG (Liquefied Natural Gas), Liquid Helium, and Liquid Oxygen in enclosed systems, ultra-low temperature freezers, blast freezers, dry ice, and in coldrooms
    Length (English) 24.75 to 27 in.
    Length (Metric) 62.5 to 68.5cm
    Material Fabrics, Multi-layered Insulated Construction
    Liner Material Thermal
    Size 11 (XL)
    Temperature (English) Min -259°F
    Color Pink
  • Description & Specifications
Related Products
Product Content Feedback