Learn More
Sklar InstrumentsSupplier Diversity Partner Sklar Black Emmett Tenaculum Hooks, Sklar, Stainless Steel with Reduced Reflectivity Polymer Coating. PK/6. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 902170
Sklar Black Emmett Tenaculum Hooks, Sklar, Stainless Steel with Reduced Reflectivity Polymer Coating. The Sklar Black Emmett Tenaculum Hook set contains 5 hooks used to hold the uterus and cervix in place during OB/GYN laser procedures. It contains one of each size/style hook that features a non-reflective black polymer coating applied over the sandblasted instrument developed specifically for laser surgeries. This safe, durable coating reduces laser reflection to less than one-tenth of a percent, ensuring a safer surgical environment for patients and surgical staff. The overall length of the instrument is 9 inches.
Catalog No. 50-143-6554