Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tenascin C Rabbit anti-Human, Clone: TNC/2981R, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP30720210UG

 View more versions of this product

Catalog No. NB121877


Add to cart

Description

Description

Tenascin C Monoclonal specifically detects Tenascin C in Human samples. It is validated for Immunohistochemistry-Paraffin.
Specifications

Specifications

Tenascin C
TNC/2981R
Unconjugated
10 mM PBS with 0.05% BSA
150-225, Cytotactin, Glioma-associated-extracellular matrix antigen, GMEM, GP 150-225, Hexabrachion, hexabrachion (tenascin C, cytotactin), hexabrachion (tenascin), HXBcytotactin, JI, MGC167029, Myotendinous antigen, neuronectin, tenascin, tenascin C, Tenascin-C, tenascin-C isoform 14/AD1/16, TN-C, TNGP
Recombinant human full-length Tenascin C protein
Protein A or G purified
RUO
Store at 4°C.
Monoclonal
Human
Immunohistochemistry (Paraffin)
0.2 mg/ml
Immunohistochemistry-Paraffin 2-4 ug/ml
Purified
Rabbit
IgG
100 μg
Breast Cancer, Cancer, Extracellular Matrix, Neuroscience, Prostate Cancer
Primary
3371
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit