Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tenascin X Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550581

Catalog No. PA550581


Add to cart

Description

Description

The antibody detects endogenous levels of total Tenascin X protein.

This gene encodes a member of the tenascin family of extracellular matrix glycoproteins. The tenascins have anti-adhesive effects, as opposed to fibronectin which is adhesive. This protein is thought to function in matrix maturation during wound healing, and its deficiency has been associated with the connective tissue disorder Ehlers-Danlos syndrome. This gene localizes to the major histocompatibility complex (MHC) class III region on chromosome 6. It is one of four genes in this cluster which have been duplicated. The duplicated copy of this gene is incomplete and is a pseudogene which is transcribed but does not encode a protein. The structure of this gene is unusual in that it overlaps the CREBL1 and CYP21A2 genes at its 5 ft. and 3 ft. ends, respectively. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications

Specifications

Tenascin X
Polyclonal
Unconjugated
TNXB
Liquid
TNXB
Synthetic peptide corresponding to a region derived from internal residues of human tenascin XB
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin)
1.5 mg/mL
PBS with 50% glycerol and 0.05% sodium azide; pH 7.3
P22105
XB, TNX, XBS, EDS3, HXBL, TENX, TN-X, TNXB1, TNXB2, TNXBS
Rabbit
IgG
100 μL
-20°C
Primary
7148
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit