Learn More
Selleck Chemical LLC
Tenofovir Disoproxil Fumarate 10mg 202138-50-9 GS-1278 Disoproxil Fumarate
Manufacturer: Selleck Chemical LLC S140010MG
Tenofovir (GS-1278) Disoproxil Fumarate belongs to a class of antiretroviral drugs, it inhibits the activity of HIV reverse transcriptase by competing with the natural substrate deoxyadenosine 5’-triphosphate and, after incorporation into DNA, by DNA chain termination.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.