Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tenovin-1, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
p53 activator that protects against MDM2-mediated p53 degradation. Elevates levels of p53 and p21CIP/WAF1 and induces expression from an endogenous p53-dependent promoter. Exhibits potent antiproliferative activity in vitro.
Specifications
Quantity | 10 mg |
Formula Weight | 369.48 |
Percent Purity | >99% |
Chemical Name or Material | Tenovin-1 |
Safety and Handling
Recommended Storage | Store at +4°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction