Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TENR Polyclonal Antibody
TENR Polyclonal Antibody
Supplier: Thermo Scientific PA593016
Description
TENR Polyclonal Antibody for Western Blot, IHC (P)

Specifications
TENR | |
Polyclonal | |
Unconjugated | |
ADAD1 | |
ADAD1; adenosine deaminase domain containing 1; adenosine deaminase domain containing 1 (testis specific); adenosine deaminase domain containing 1 (testis-specific); adenosine deaminase domain-containing protein 1; TENR; testis nuclear RNA binding protein; testis nuclear RNA-binding protein; testis tissue sperm-binding protein Li 92mP | |
Rabbit | |
Affinity Chromatography | |
RUO | |
132612, 294979 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.65 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q3KR54, Q96M93 | |
ADAD1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human ADAD1 (NP_0011527571). | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction