Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TentaGel HL-RAM, ACROS Organics
Specifications
Chemical Name or Material | TentaGel™ HL-RAM |
---|---|
MDL Number | MFCD00217896 |
Synonym | N-FMOC-4'-[poly(oxyethylene)carbamoylmethoxy]-2, 4-dimethoxy-benzhydrylamine polymer bound, Poly(oxyethylene)-RAM polymer bound |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
AC354460010
|
Acros Organics
354460010 |
1g | Glass bottle |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
AC354460050
|
Acros Organics
354460050 |
5g | Glass bottle |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
Description
For solid support synthesisSpecifications
TentaGel™ HL-RAM | |
MFCD00217896 | |
N-FMOC-4'-[poly(oxyethylene)carbamoylmethoxy]-2, 4-dimethoxy-benzhydrylamine polymer bound, Poly(oxyethylene)-RAM polymer bound |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title