Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TentaGel MB-Br, ACROS Organics
$76.00 - $311.18
Specifications
Chemical Name or Material | TentaGel™ MB-Br |
---|---|
MDL Number | MFCD00217898 |
Synonym | O-(2-Bromoethyl)polyethylene glycol polymer bound |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
AC354480050
|
Acros Organics
354480050 |
5g | Glass bottle |
Each for $311.18
Order in Bulk
Request bulk or custom formats |
|
AC354480010
|
Acros Organics
354480010 |
1g | Glass bottle |
Each for $76.00
Order in Bulk
Request bulk or custom formats |
|
Description
For solid support synthesisSpecifications
TentaGel™ MB-Br | |
MFCD00217898 | |
O-(2-Bromoethyl)polyethylene glycol polymer bound |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title