Learn More
TentaGel MB-CH(OEt)2, ACROS Organics
Manufacturer: Acros Organics 354490050
Description
For solid support synthesisSpecifications
TentaGel™ MB-CH(OEt)2 | |
Glass bottle | |
5g | |
MFCD01868634 | |
Powder |
0.4 to 0.6mmol/g | |
140 to 170 Micron | |
White to Yellow | |
O-[2-(6, 6-Diethoxyhexanoylamino)ethyl]polyethylene glycol polymer bound |