Learn More
TentaGel™ MB-OH, ACROS Organics™
Manufacturer:  Acros Organics 354520010
 View more versions of this product
Catalog No. AC354520010
Order in Bulk
Request bulk or custom formats
Add to cart
Description
Description
For solid support synthesis
Specifications
Specifications
TentaGel™ MB-OH
Glass bottle
1g
MFCD00217900
Powder
0.4 to 0.6mmol/g
140 to 170 Micron
White to Yellow
O-(2-Hydroxyethyl)polyethylene glycol polymer bound