Learn More
TentaGel™ MB-OH, ACROS Organics™
Manufacturer:  Acros Organics 354520050
 View more versions of this product
Catalog No. AC354520050
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
For solid support synthesis
Specifications
Specifications
TentaGel™ MB-OH
Glass bottle
5g
MFCD00217900
Powder
0.4 to 0.6mmol/g
140 to 170 Micron
White to Yellow
O-(2-Hydroxyethyl)polyethylene glycol polymer bound