Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Teprotumumab, Mouse anti-Many, Alexa Fluor 350, Clone: 975547, R&D Systems™
Mouse Monoclonal Antibody
Supplier: R&D Systems FAB96341U100UG
This item is not returnable.
View return policy
Description
Teprotumumab Monoclonal specifically detects Teprotumumab in Multi-Species samples. It is validated for Flow Cytometry.Specifications
Teprotumumab | |
Monoclonal | |
Alexa Fluor 350 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
Full-length Teprotumumab | |
100 μg | |
Primary | |
Many | |
Purified |
Flow Cytometry | |
975547 | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Detects Teprotumumab in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG2a |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title