Learn More
Teprotumumab, Mouse anti-Many, Alexa Fluor 594, Clone: 975547, R&D Systems™
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB96341T100UG
Catalog No. FAB96341T10
Add to cart
This item is not returnable. View return policy
Description
Description
Teprotumumab Monoclonal antibody specifically detects Teprotumumab in Multi-Species Samples. It is validated for Flow Cytometry.
Specifications
Specifications
Teprotumumab
975547
Flow Cytometry 0.25-1 ug/10^6 cells
Purified
Full-length Teprotumumab
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Detects Teprotumumab in direct ELISAs.
Flow Cytometry
Alexa Fluor 594
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Mouse
IgG2a
100 ug
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Monoclonal
Many