Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TER-119/Erythroid Cells Rat anti-Mouse, R718, Clone: TER-119, BD Horizon™
Rat Monoclonal Antibody
Supplier: BD Biosciences 567363
Description
TER-119/Erythroid Cells Monoclonal antibody specifically detects TER-119/Erythroid Cells in Mouse samples. It is validated for Flow CytometrySpecifications
TER-119/Erythroid Cells | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed. | |
IgG2b κ |
Flow Cytometry | |
TER-119 | |
R718 | |
Lymphocyte antigen 76; Ly76; Ly-76; TER-119; Ter119 | |
Mouse Fetal Liver | |
RUO | |
Mouse | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title