Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Terazosin Hydrochloride, Dihydrate, USP, Spectrum™

Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Amber Glass Bottle
70024-40-7
C19H30ClN5O6
459.93
NZMOFYDMGFQZLS-UHFFFAOYNA-N
hydrogen 6,7-dimethoxy-2-[4-(oxolane-2-carbonyl)piperazin-1-yl]quinazolin-4-amine dihydrate chloride
[H+].O.O.[Cl-].COC1=C(OC)C=C2C(N)=NC(=NC2=C1)N1CCN(CC1)C(=O)C1CCCO1
70024-40-7
C19H30ClN5O6
459.93
NZMOFYDMGFQZLS-UHFFFAOYNA-N
hydrogen 6,7-dimethoxy-2-[4-(oxolane-2-carbonyl)piperazin-1-yl]quinazolin-4-amine dihydrate chloride
[H+].O.O.[Cl-].COC1=C(OC)C=C2C(N)=NC(=NC2=C1)N1CCN(CC1)C(=O)C1CCCO1

T3019, 70024-40-7, C19H25N5O4•HCl•2H2O

Spectrum™ Terazosin Hydrochloride, Dihydrate, USP is in a group of drugs called alpha-adrenergic blockers that are used to treat hypertension. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Amber Glass Bottle
Quantity 500 g
CAS Max % 1
Grade USP