Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Terbinafine Hydrochloride, EP, 99-101%, Spectrum™

Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
78628-80-5
C21H26ClN
327.90
BWMISRWJRUSYEX-SZKNIZGXSA-N
hydrogen [(2E)-6,6-dimethylhept-2-en-4-yn-1-yl](methyl)[(naphthalen-1-yl)methyl]amine chloride
[H+].[Cl-].CN(C\C=C\C#CC(C)(C)C)CC1=C2C=CC=CC2=CC=C1
78628-80-5
C21H26ClN
327.90
BWMISRWJRUSYEX-SZKNIZGXSA-N
hydrogen [(2E)-6,6-dimethylhept-2-en-4-yn-1-yl](methyl)[(naphthalen-1-yl)methyl]amine chloride
[H+].[Cl-].CN(C\C=C\C#CC(C)(C)C)CC1=C2C=CC=CC2=CC=C1

T3880, 78628-80-5, C21H25N•HCl

Spectrum™ Terbinafine Hydrochloride, EP is used to treat certain types of fungal infections (e.g., fingernail or toenail). The EP grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications. All Spectrum Chemical EP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Amber Glass Bottle
Loss on Drying 0.005
Quantity 5 g
CAS Max % 1
Percent Purity 99 to 101%