Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Terbium Foil, 0.025mm (0.001 in.) thick, (REO), 0.999, Alfa Aesar™

Form: Approx. 0.13g/25 x 25mm

Manufacturer:  Alfa Aesar 12431FI

Quantity 50 x 50mm

 View more versions of this product

Catalog No. AA12431FI

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

7440-27-9
99.9% (REO)
Terbium
Foil, 0.025mm (0.001 in.) thick
MFCD00011256

Specifications

7440-27-9
99.9% (REO)
MFCD00011256
50 x 50mm
Foil, 0.025mm (0.001 in.) thick
Silver Gray foil
Silver-gray
Terbium

Safety and Handling

SDS & Safety Documents

H228-H260; Flammable solid. In contact with water releases flammable gases which may ignite spontaneously.

DOTInformation : DOT Class: 4.1, Packaging Group: II

EINECSNumber : 231-137-6

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit