Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Terbium foil, 0.62mm (0.024in) thick, 99.9% (REO), Thermo Scientific™

Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
7440-27-9
Tb
158.925
MFCD00011256
GZCRRIHWUXGPOV-UHFFFAOYSA-N
unii-06ssf7p179, ingot, foil, terbio, atom, chips, pieces, powder, acmc-20ajly, pieces, distilled dendritic
23958
CHEBI:33376
terbium
[Tb]
7440-27-9
Tb
158.925
MFCD00011256
GZCRRIHWUXGPOV-UHFFFAOYSA-N
unii-06ssf7p179, ingot, foil, terbio, atom, chips, pieces, powder, acmc-20ajly, pieces, distilled dendritic
23958
CHEBI:33376
terbium
[Tb]
  • Packaged under argon
Quantity 50 x 50 mm
Weight ≈3.2g/25x25mm
Packaging Mylar under Argon
Physical Form Foil
Percent Purity 99.9%
Chemical Name or Material Terbium foil, 0.62mm (0.024 in.) thick
Assay (REO)

EINECSNumber : 231-137-6

TSCA : Yes

storageNote1 :

  • Packaged under argon

Recommended Storage : Ambient temperatures

RUO – Research Use Only