Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Terbium(III) 2,4-pentanedionate, 99.9% (REO)
Click to view available options
Quantity:
2g
10g
Specifications
Solubility Information | Insoluble in water. |
Formula Weight | 456.25 |
Physical Form | Powder |
Percent Purity | 99.9% |
Odor | Odorless |
Quantity | 2g |
Chemical Name or Material | Terbium(III) 2,4-pentanedionate |
Assay | (REO) |
Safety and Handling
EINECSNumber : 238-195-1
TSCA : No
Recommended Storage : Ambient temperatures