Terbium(III) acetate hydrate, 99.9% (REO), Alfa Aesar™, Reacton™

Manufacturer: Alfa Aesar 1460009

Quantity 10g

 View more versions of this product

Catalog No. AA1460009

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 100587-92-6
    Physical Form Crystalline Aggregates
    Assay Percent Range 99.9% (REO)
    Molecular Formula Tb(OOCCH3)3·xH2O
    Formula Weight 336.08 (anhydrous)
    MDL Number MFCD00149233
    Chemical Name or Material Terbium(III) acetate hydrate

    Specifications

    Physical Form Crystalline Aggregates
    Formula Weight 336.08 (anhydrous)
    Quantity 10g
    Assay Percent Range 99.9% (REO)
    Molecular Formula Tb(OOCCH3)3·xH2O
    MDL Number MFCD00149233
    CAS 100587-92-6
    Chemical Name or Material Terbium(III) acetate hydrate

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit