Alfa Aesar™ Terbium(III,IV) oxide, 99.9% (REO)

Manufacturer: Alfa Aesar™ 3624604

Quantity 2g

 View more versions of this product

Catalog No. AA3624604

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 12037-01-3
    Formula Weight 747.69
    MDL Number MFCD00083151
    Merck Index 14,9157
    Chemical Name or Material Terbium(III,IV) oxide

    Specifications

    Formula Weight 747.69
    Odor Odorless
    Physical Form Sintered Lump
    MDL Number MFCD00083151
    Merck Index 14,9157
    TSCA TSCA
    Particle Size 3 to 12mm
    CAS 12037-01-3
    Melting Point 2340°C
    Quantity 2g
    Chemical Name or Material Terbium(III,IV) oxide
    Assay 99.9%, (REO)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit