Learn More
Terbium(III,IV) oxide, REacton™, 99.9% (REO), Thermo Scientific Chemicals
Description
Terbium(III,IV) oxide acts as a precursor for the preparation of other terbium compounds. It is used as a redox catalyst in reactions involving oxygen. At high temperatures, it can liberate oxygen. It finds applications in the manufacture of glass, optical and ceramic. Nanoparticles of terbium oxide are used as analytical reagents for the determination of drugs in food.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 2340°C |
Physical Form | Powder |
Quantity | 10 g |
Assay Percent Range | 99.9% (REO) |
Solubility Information | Soluble in dilute acids. Insoluble in water. |
Formula Weight | 747.69 |
Chemical Name or Material | Terbium(III, IV) oxide |
Safety and Handling
EINECSNumber | 234-856-3 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.