Learn More
Terbium(III,IV) oxide, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Description
Terbium(III,IV) oxide is used in the preparation of other terbium compounds. Activating agents for fluorescent powders, composites for catalytic automobile exhaust converters, Glass materials with a Faraday Rotation effect for optical and laser-based devices, Magneto-optical glasses, Magneto-optical recording materials.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 2340°C |
Physical Form | Powder |
Quantity | 25 g |
Assay Percent Range | 99.99% (REO) |
Solubility Information | soluble in dilute acids. Insoluble in water. |
Formula Weight | 747.69 |
Chemical Name or Material | Terbium(III, IV) oxide |
Safety and Handling
EINECSNumber | 234-856-3 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.