Alfa Aesar™ Terbium(III) perchlorate, 50% w/w aq. soln., Reagent Grade

Manufacturer: Alfa Aesar™ 40573NA

Quantity 2g

 View more versions of this product

Catalog No. AA40573NA

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 14014-09-6
    Linear Formula Tb(ClO4)3·6H2O
    Formula Weight 565.37 (457.28 Anhydrous)
    MDL Number MFCD00150362
    UN Number UN3211
    Chemical Name or Material Terbium(III) perchlorate
    Grade Reagent Grade

    Specifications

    Formula Weight 565.37 (457.28 Anhydrous)
    Health Hazard 3 GHS P Statement
    P221-P210-P220-P305+P351+P338-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Keep/Store away from clothing/combustible materials.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00150362
    CAS 14014-09-6
    Health Hazard 1 Danger
    Density 1.5g/mL
    DOT Information Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, AQUEOUS SOLUTION, N.O.S.
    Odor Odorless
    Linear Formula Tb(ClO4)3·6H2O
    UN Number UN3211
    Quantity 2g
    Concentration 50% w/w aq. soln.
    Physical Form Liquid
    Color Colorless
    Health Hazard 2 GHS H Statement
    H272-H315-H319-H335
    May intensify fire
    oxidizer.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Chemical Name or Material Terbium(III) perchlorate
    Grade Reagent Grade

    GHS H Statement
    H272-H315-H319-H335
    May intensify fire
    oxidizer.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P221-P210-P220-P305+P351+P338-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Keep/Store away from clothing/combustible materials.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit