Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Terbium pieces, distilled dendritic, 99.9% (REO)

Click to view available options
Quantity:
1 g
5 g
25 g
7440-27-9
Tb
158.925
MFCD00011256
GZCRRIHWUXGPOV-UHFFFAOYSA-N
unii-06ssf7p179, ingot, foil, terbio, atom, chips, pieces, powder, acmc-20ajly, pieces, distilled dendritic
23958
CHEBI:33376
terbium
[Tb]
7440-27-9
Tb
158.925
MFCD00011256
GZCRRIHWUXGPOV-UHFFFAOYSA-N
unii-06ssf7p179, ingot, foil, terbio, atom, chips, pieces, powder, acmc-20ajly, pieces, distilled dendritic
23958
CHEBI:33376
terbium
[Tb]
  • Packaged under argon
Quantity 1 g
UN Number UN3178
Odor Odorless
Physical Form Pieces
Percent Purity 99.9%
Color Silver to Gray
Chemical Name or Material Terbium pieces, Distilled dendritic
Assay (REO)

GHS H Statement
H228-H260
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.

P210-P235+P410-P240-P241-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q

H228-H315-H319-X

DOTInformation : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.

EINECSNumber : 231-137-6

TSCA : Yes

storageNote1 :

  • Packaged under argon

Recommended Storage : Ambient temperatures