Terbium Pieces, (REO), 0.999, Alfa Aesar™

Manufacturer: Alfa Aesar 0032406

Quantity 5g

 View more versions of this product

Catalog No. AA0032406

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-27-9
    Physical Form Pieces
    Assay Percent Range 99.9% (REO)
    MDL Number MFCD00011256
    Chemical Name or Material Terbium

    Specifications

    Quantity 5g
    Color Silver-gray
    DOT Information DOT Class: 4.1, Packaging Group: II
    EINECS Number 231-137-6
    Physical Form Pieces
    Odor Odorless
    Assay Percent Range 99.9% (REO)
    Appearance Silver Gray pieces
    MDL Number MFCD00011256
    CAS 7440-27-9
    Health Hazard 1 H228-H260; Flammable solid. In contact with water releases flammable gases which may ignite spontaneously.
    Chemical Name or Material Terbium

    H228-H260; Flammable solid. In contact with water releases flammable gases which may ignite spontaneously.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit