Alfa Aesar™ Terbium powder, -40 mesh, 99.9% (REO)

Manufacturer: Alfa Aesar™ 0032206

Quantity 5g

 View more versions of this product

Catalog No. AA0032206

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-27-9
    MDL Number MFCD00011256
    UN Number UN3089
    Chemical Name or Material Terbium powder

    Specifications

    Quantity 5g
    Health Hazard 3 GHS P Statement
    P210-P231+P232-P280-P370+P378b-P402+P404-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Handle under inert gas.
    Protect from moisture.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: Special powder for metal fires.
    Store in a dry place.
    Store in a closed container.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Packaging Poly bottle
    MDL Number MFCD00011256
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.
    Physical Form Powder
    UN Number UN3089
    Mesh Size −40 Mesh
    Sensitivity Air & Moisture Sensitive
    Color Silver-gray
    Odor Odorless
    TSCA TSCA
    CAS 7440-27-9
    Health Hazard 2 GHS H Statement
    H228-H260
    Flammable solid.
    In contact with water releases flammable gases which may ignite spontaneously.
    Storage Note 1
    • Packaged in air stable poly bottle
    Chemical Name or Material Terbium powder
    Assay 99.9%, (REO)

    GHS H Statement
    H228-H260
    Flammable solid.
    In contact with water releases flammable gases which may ignite spontaneously.

    GHS P Statement
    P210-P231+P232-P280-P370+P378b-P402+P404-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Handle under inert gas.
    Protect from moisture.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: Special powder for metal fires.
    Store in a dry place.
    Store in a closed container.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    • Packaged in air stable poly bottle

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit