Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Terbium sputtering target, 50.8mm dia x 1.59mm thick, Thermo Scientific™

Click to view available options
Quantity:
1 ea.
7440-27-9
Tb
158.925
MFCD00011256
GZCRRIHWUXGPOV-UHFFFAOYSA-N
unii-06ssf7p179, ingot, foil, terbio, atom, chips, pieces, powder, acmc-20ajly, pieces, distilled dendritic
23958
CHEBI:33376
terbium
[Tb]
7440-27-9
Tb
158.925
MFCD00011256
GZCRRIHWUXGPOV-UHFFFAOYSA-N
unii-06ssf7p179, ingot, foil, terbio, atom, chips, pieces, powder, acmc-20ajly, pieces, distilled dendritic
23958
CHEBI:33376
terbium
[Tb]
Quantity 1 ea.
Chemical Name or Material Terbium sputtering target, 50.8mm dia. x 1.59mm thick

EINECSNumber : 231-137-6

TSCA : Yes

Recommended Storage : Ambient temperatures