Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Terephthalaldehyde mono(diethyl acetal), 97%, stab., Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
4-(Diethoxymethyl)benzaldehyde was used in the synthesis of 4-(diethoxymethyl)benzyl alcohol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-(Diethoxymethyl)benzaldehyde was used in the synthesis of 4-(diethoxymethyl)benzyl alcohol.
Solubility
soluble in alcohol, chlorinated solvents and toluene. Decomposes in water.
Notes
Air sensitive. Store under inert gas. Keep way from oxidizing agents, air.
4-(Diethoxymethyl)benzaldehyde was used in the synthesis of 4-(diethoxymethyl)benzyl alcohol.
Solubility
soluble in alcohol, chlorinated solvents and toluene. Decomposes in water.
Notes
Air sensitive. Store under inert gas. Keep way from oxidizing agents, air.
Specifications
Density | 1.047 |
Boiling Point | 89°C to 93°C (7 mmHg) |
Flash Point | >110°C (230°F) |
Refractive Index | 1.507 |
Quantity | 25 g |
Beilstein | 4293872 |
Sensitivity | Air Sensitive |
Solubility Information | soluble in alcohol,chlorinated solvents and toluene. Decomposes in water. |
Formula Weight | 208.26 |
Percent Purity | 98% |
Show More |
Safety and Handling
Hazard Category | H315-H319-H335 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
EINECSNumber | 000-000-0 |
TSCA | No |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction