Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Terephthalaldehyde mono(diethyl acetal), 98%, stab.

Click to view available options
Quantity:
5g
25g
100g
500g
81172-89-6
C12H16O3
208.257
MFCD00010217
HTMXMFARWHNJDW-UHFFFAOYSA-N
4-diethoxymethyl benzaldehyde,terephthalaldehyde mono diethyl acetal,benzaldehyde, 4-diethoxymethyl,terephthaldehyde mono diethyl acetal,terephthalaldehyde mono-diethyl acetal,pubchem22621,acmc-209tbs,4-diethoxymethyl-benzaldehyde,4-diethoxymethy benzaldehyde,4-diethyoxymethyl benzaldehyde
595993
4-(diethoxymethyl)benzaldehyde
CCOC(C1=CC=C(C=C1)C=O)OCC
81172-89-6
C12H16O3
208.257
MFCD00010217
HTMXMFARWHNJDW-UHFFFAOYSA-N
4-diethoxymethyl benzaldehyde,terephthalaldehyde mono diethyl acetal,benzaldehyde, 4-diethoxymethyl,terephthaldehyde mono diethyl acetal,terephthalaldehyde mono-diethyl acetal,pubchem22621,acmc-209tbs,4-diethoxymethyl-benzaldehyde,4-diethoxymethy benzaldehyde,4-diethyoxymethyl benzaldehyde
595993
4-(diethoxymethyl)benzaldehyde
CCOC(C1=CC=C(C=C1)C=O)OCC
Applications
4-(Diethoxymethyl)benzaldehyde was used in the synthesis of 4-(diethoxymethyl)benzyl alcohol.

Solubility
soluble in alcohol, chlorinated solvents and toluene. Decomposes in water.

Notes
Air sensitive. Store under inert gas. Keep way from oxidizing agents, air.
Refractive Index 1.507
Quantity 100g
Boiling Point 89°C to 93°C (7mmHg)
Density 1.047
Sensitivity Air Sensitive
Flash Point >110°C (230°F)
Beilstein 4293872
Solubility Information soluble in alcohol,chlorinated solvents and toluene. Decomposes in water.
Formula Weight 208.26
Percent Purity 98%
Chemical Name or Material Terephthalaldehyde mono(diethyl acetal), stabilized
Show More Show Less

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen